Assay Details
| Gene Symbol: PYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337783.1 | NP_001326563.1 | 3 - 4 | 818 | 71 | |
| NM_111720.4 | NP_187498.1 | 3 - 4 | 981 | 71 | ||
| GenBank mRNA | AK226401.1 | - | 3 - 4 | 719 | 71 | |
| AY099816.1 | - | 3 - 4 | 718 | 71 | ||
More Information
Set Membership: |
|

PYD4
Genomic Map