Assay Details
| Gene Symbol: PYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337783.1 | NP_001326563.1 | 5 - 6 | 1145 | 73 | |
| NM_111720.4 | NP_187498.1 | 5 - 6 | 1308 | 73 | ||
| GenBank mRNA | AK226401.1 | - | 5 - 6 | 1046 | 73 | |
| AY099816.1 | - | 5 - 6 | 1045 | 73 | ||
More Information
Set Membership: |
|

PYD4
Genomic Map