Assay Details
| Gene Symbol: HCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111729.3 | NP_566339.1 | 3 - 4 | 549 | 88 | |
| GenBank mRNA | AY070393.1 | - | 3 - 4 | 527 | 88 | |
| AY085259.1 | - | 3 - 4 | 527 | 88 | ||
| AY096736.1 | - | 3 - 4 | 457 | 88 | ||
More Information
Set Membership: |
|

HCC1
Genomic Map