Assay Details
Gene Symbol: HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111729.3 | NP_566339.1 | 5 - 6 | 857 | 71 | |
GenBank mRNA | AY070393.1 | - | 5 - 6 | 835 | 71 | |
AY085259.1 | - | 5 - 6 | 71 | |||
AY096736.1 | - | 5 - 6 | 765 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
