Assay Details
| Gene Symbol: HCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111729.3 | NP_566339.1 | 6 - 7 | 989 | 117 | |
| GenBank mRNA | AY070393.1 | - | 6 - 7 | 967 | 117 | |
| AY085259.1 | - | 6 - 7 | 967 | 117 | ||
| AY096736.1 | - | 6 - 7 | 897 | 117 | ||
More Information
Set Membership: |
|

HCC1
Genomic Map