Assay Details
| Gene Symbol: PYK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111760.3 | NP_187537.1 | 2 - 3 | 279 | 66 | |
| GenBank mRNA | AF386967.1 | - | 2 - 3 | 273 | 66 | |
| AK226844.1 | - | 277 | 66 | |||
| AK230345.1 | - | 1038 | 66 | |||
| AK317443.1 | - | 2 - 3 | 276 | 66 | ||
| AY136440.1 | - | 2 - 3 | 273 | 66 | ||
| AY140060.1 | - | 2 - 3 | 276 | 66 | ||
| BT000230.1 | - | 2 - 3 | 228 | 66 | ||
| Gene Symbol: AT4G02740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK228382.1 | - | 954 | 66 | ||
More Information
Set Membership: |
|

PYK10
Genomic Map