Assay Details
| Gene Symbol: Fes1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084657.1 | NP_001078126.1 | 4 - 5 | 499 | 64 | |
| NM_001161128.1 | NP_001154600.1 | 3 - 4 | 743 | 64 | ||
| NM_001337811.1 | NP_001327344.1 | 4 - 5 | 600 | 64 | ||
| NM_111769.2 | NP_566349.1 | 3 - 4 | 518 | 64 | ||
| GenBank mRNA | BT004332.1 | - | 3 - 4 | 355 | 64 | |
| BT005589.1 | - | 3 - 4 | 272 | 64 | ||
More Information
Set Membership: |
|

Fes1A
Genomic Map