Assay Details
| Gene Symbol: Fes1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084657.1 | NP_001078126.1 | 5 - 6 | 720 | 76 | |
| NM_001161128.1 | NP_001154600.1 | 4 - 5 | 964 | 76 | ||
| NM_001337811.1 | NP_001327344.1 | 5 - 6 | 821 | 76 | ||
| NM_111769.2 | NP_566349.1 | 4 - 5 | 739 | 76 | ||
| GenBank mRNA | BT004332.1 | - | 4 - 5 | 576 | 76 | |
| BT005589.1 | - | 4 - 5 | 493 | 76 | ||
More Information
Set Membership: |
|

Fes1A
Genomic Map