Assay Details
| Gene Symbol: MYB3R-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084658.2 | NP_001078127.1 | 4 - 5 | 510 | 81 | |
| NM_001337814.1 | NP_001327447.1 | 3 - 4 | 818 | 81 | ||
| NM_111771.4 | NP_566350.1 | 4 - 5 | 495 | 81 | ||
| GenBank mRNA | AY142649.1 | - | 4 - 5 | 314 | 81 | |
| AY519651.1 | - | 4 - 5 | 314 | 81 | ||
More Information
Set Membership: |
|

MYB3R-3
Genomic Map