Assay Details
| Gene Symbol: MYB3R-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084658.2 | NP_001078127.1 | 7 - 8 | 839 | 128 | |
| NM_001337814.1 | NP_001327447.1 | 6 - 7 | 1147 | 128 | ||
| NM_111771.4 | NP_566350.1 | 7 - 8 | 824 | 128 | ||
| GenBank mRNA | AY142649.1 | - | 7 - 8 | 643 | 128 | |
| AY519651.1 | - | 7 - 8 | 643 | 128 | ||
More Information
Set Membership: |
|

MYB3R-3
Genomic Map