Assay Details
| Gene Symbol: PLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202917.1 | NP_001189846.1 | 1 - 2 | 1549 | 135 | |
| NM_111774.2 | NP_187551.1 | 1 - 2 | 1532 | 135 | ||
More Information
Set Membership: |
|

PLL3
Genomic Map