Assay Details
| Gene Symbol: PAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125132.1 | NP_001118604.1 | 2 - 3 | 1879 | 75 | |
| NM_001337827.1 | NP_001326239.1 | 1 - 2 | 2260 | 75 | ||
| NM_111790.4 | NP_187567.1 | 2 - 3 | 1973 | 75 | ||
| NM_202539.2 | NP_974268.1 | 2 - 3 | 1872 | 75 | ||
More Information
Set Membership: |
|

PAH1
Genomic Map