Assay Details
| Gene Symbol: RVE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035585.4 | NP_001030662.1 | 1 - 2 | 401 | 105 | |
| NM_001337829.1 | NP_001327351.1 | 1 - 2 | 401 | 105 | ||
| NM_001337831.1 | NP_001327349.1 | 1 - 2 | 345 | 105 | ||
| NM_001337832.1 | NP_001327348.1 | 1 - 2 | 345 | 105 | ||
| NM_111794.4 | NP_187571.2 | 1 - 2 | 401 | 105 | ||
| GenBank mRNA | AJ937213.1 | - | 1 - 2 | 181 | 105 | |
| AY091106.1 | - | 1 - 2 | 225 | 105 | ||
| AY122969.1 | - | 1 - 2 | 181 | 105 | ||
| AY550307.1 | - | 1 - 2 | 181 | 105 | ||
More Information
Set Membership: |
|

RVE8
Genomic Map