Assay Details
| Gene Symbol: RVE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035585.4 | NP_001030662.1 | 8 - 9 | 1161 | 113 | |
| NM_001337828.1 | NP_001327347.1 | 7 - 8 | 1426 | 113 | ||
| NM_001337829.1 | NP_001327351.1 | 7 - 8 | 1333 | 113 | ||
| NM_001337830.1 | NP_001327350.1 | 7 - 8 | 1392 | 113 | ||
| NM_111794.4 | NP_187571.2 | 8 - 9 | 1139 | 113 | ||
| GenBank mRNA | AY091106.1 | - | 8 - 9 | 963 | 113 | |
More Information
Set Membership: |
|

RVE8
Genomic Map