Assay Details
| Gene Symbol: AT3G09720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337839.1 | NP_001326599.1 | 11 - 12 | 1446 | 71 | |
| NM_111806.7 | NP_187583.2 | 11 - 12 | 1417 | 71 | ||
| GenBank mRNA | AK227326.1 | - | 11 - 12 | 1334 | 71 | |
| BT005833.1 | - | 10 - 11 | 1256 | 71 | ||
More Information
Set Membership: |
|

AT3G09720
Genomic Map