Assay Details
| Gene Symbol: AT3G09720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337839.1 | NP_001326599.1 | 7 - 8 | 804 | 82 | |
| NM_111806.7 | NP_187583.2 | 7 - 8 | 775 | 82 | ||
| GenBank mRNA | AK227326.1 | - | 7 - 8 | 692 | 82 | |
| BT005833.1 | - | 6 - 7 | 614 | 82 | ||
More Information
Set Membership: |
|

AT3G09720
Genomic Map