Assay Details
| Gene Symbol: SYP71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111809.5 | NP_566354.1 | 1 - 2 | 421 | 100 | |
| GenBank mRNA | AF355757.1 | - | 1 - 2 | 199 | 100 | |
| AY090358.1 | - | 1 - 2 | 267 | 100 | ||
| BT024483.1 | - | 1 - 2 | 162 | 100 | ||
More Information
Set Membership: |
|

SYP71
Genomic Map