Assay Details
| Gene Symbol: SYP71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111809.5 | NP_566354.1 | 8 - 9 | 1040 | 103 | |
| GenBank mRNA | AF355757.1 | - | 8 - 9 | 818 | 103 | |
| AY090358.1 | - | 8 - 9 | 886 | 103 | ||
More Information
Set Membership: |
|

SYP71
Genomic Map