Assay Details
| Gene Symbol: AT3G10060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111841.5 | NP_187617.1 | 6 - 7 | 674 | 65 | |
| GenBank mRNA | AY094442.1 | - | 6 - 7 | 524 | 65 | |
| AY122895.1 | - | 6 - 7 | 457 | 65 | ||
More Information
Set Membership: |
|

AT3G10060
Genomic Map