Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111842.6 | NP_566367.1 | 1 - 2 | 600 | 67 | |
| GenBank mRNA | AF344878.1 | - | 1 - 2 | 572 | 67 | |
| AK221690.1 | - | 1 - 2 | 589 | 67 | ||
| AY463613.1 | - | 1 - 2 | 572 | 67 | ||
More Information
Set Membership: |
|

TAF12
Genomic Map