Assay Details
| Gene Symbol: AT3G10440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111879.3 | NP_187655.2 | 15 - 16 | 1295 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G10440| Gene Symbol: AT3G10440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111879.3 | NP_187655.2 | 15 - 16 | 1295 | 73 | |
Set Membership: |
|