Assay Details
| Gene Symbol: AT3G10440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337875.1 | NP_001326768.1 | 1 - 2 | 91 | 91 | |
| NM_111879.3 | NP_187655.2 | 1 - 2 | 91 | 91 | ||
More Information
Set Membership: |
|

AT3G10440
Genomic Map