Assay Details
| Gene Symbol: HB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111887.3 | NP_187663.1 | 1 - 2 | 268 | 84 | |
| GenBank mRNA | AY087650.1 | - | 1 - 2 | 198 | 84 | |
| BT028909.1 | - | 1 - 2 | 103 | 84 | ||
More Information
Set Membership: |
|

HB2
Genomic Map