Assay Details
| Gene Symbol: HB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111887.3 | NP_187663.1 | 3 - 4 | 499 | 78 | |
| GenBank mRNA | AY087650.1 | - | 3 - 4 | 429 | 78 | |
| BT028909.1 | - | 3 - 4 | 334 | 78 | ||
More Information
Set Membership: |
|

HB2
Genomic Map