Assay Details
| Gene Symbol: NAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111903.3 | NP_187678.1 | 1 - 2 | 503 | 60 | |
| GenBank mRNA | AY035125.1 | - | 1 - 2 | 444 | 60 | |
| AY113917.1 | - | 1 - 2 | 361 | 60 | ||
More Information
Set Membership: |
|

NAP7
Genomic Map