Assay Details
| Gene Symbol: NAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111903.3 | NP_187678.1 | 4 - 5 | 898 | 69 | |
| GenBank mRNA | AY035125.1 | - | 4 - 5 | 839 | 69 | |
| AY113917.1 | - | 4 - 5 | 756 | 69 | ||
More Information
Set Membership: |
|

NAP7
Genomic Map