Assay Details
| Gene Symbol: NAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111903.3 | NP_187678.1 | 5 - 6 | 1075 | 95 | |
| GenBank mRNA | AY035125.1 | - | 5 - 6 | 1016 | 95 | |
| AY113917.1 | - | 5 - 6 | 933 | 95 | ||
More Information
Set Membership: |
|

NAP7
Genomic Map