Assay Details
| Gene Symbol: ASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337892.1 | NP_001326565.1 | 12 - 13 | 2114 | 91 | |
| NM_001337893.1 | NP_001326566.1 | 12 - 13 | 2263 | 91 | ||
| NM_001337894.1 | NP_001326567.1 | 11 - 12 | 1388 | 91 | ||
| NM_111911.5 | NP_187685.1 | 12 - 13 | 1926 | 91 | ||
More Information
Set Membership: |
|

ASD1
Genomic Map