Assay Details
| Gene Symbol: ASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337892.1 | NP_001326565.1 | 17 - 18 | 2695 | 112 | |
| NM_001337893.1 | NP_001326566.1 | 17 - 18 | 2844 | 112 | ||
| NM_001337894.1 | NP_001326567.1 | 16 - 17 | 1969 | 112 | ||
| NM_111911.5 | NP_187685.1 | 17 - 18 | 2507 | 112 | ||
More Information
Set Membership: |
|

ASD1
Genomic Map