Assay Details
| Gene Symbol: ASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337892.1 | NP_001326565.1 | 3 - 4 | 1067 | 70 | |
| NM_001337893.1 | NP_001326566.1 | 3 - 4 | 1216 | 70 | ||
| NM_001337894.1 | NP_001326567.1 | 2 - 3 | 341 | 70 | ||
| NM_111911.5 | NP_187685.1 | 3 - 4 | 879 | 70 | ||
More Information
Set Membership: |
|

ASD1
Genomic Map