Assay Details
| Gene Symbol: FER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111942.4 | NP_187716.1 | 7 - 8 | 892 | 95 | |
| GenBank mRNA | AJ312192.1 | - | 7 - 8 | 751 | 95 | |
| AK175807.1 | - | 7 - 8 | 772 | 95 | ||
| AK175899.1 | - | 7 - 8 | 772 | 95 | ||
| AK175901.1 | - | 7 - 8 | 772 | 95 | ||
| AK175910.1 | - | 7 - 8 | 772 | 95 | ||
More Information
Set Membership: |
|

FER2
Genomic Map