Assay Details
| Gene Symbol: FAD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111953.3 | NP_187727.1 | 5 - 6 | 1346 | 73 | |
| GenBank mRNA | D26019.1 | - | 5 - 6 | 1055 | 73 | |
| L22961.1 | - | 5 - 6 | 933 | 73 | ||
More Information
Set Membership: |
|

FAD7
Genomic Map