Assay Details
| Gene Symbol: PP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111974.4 | NP_187748.1 | 1 - 2 | 646 | 114 | |
| GenBank mRNA | AY074368.1 | - | 1 - 2 | 586 | 114 | |
| AY091391.1 | - | 1 - 2 | 445 | 114 | ||
| D38109.1 | - | 1 - 2 | 522 | 114 | ||
More Information
Set Membership: |
|

PP2CA
Genomic Map