Assay Details
| Gene Symbol: PP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111974.4 | NP_187748.1 | 2 - 3 | 983 | 66 | |
| GenBank mRNA | AY074368.1 | - | 2 - 3 | 923 | 66 | |
| AY091391.1 | - | 2 - 3 | 782 | 66 | ||
| D38109.1 | - | 2 - 3 | 859 | 66 | ||
More Information
Set Membership: |
|

PP2CA
Genomic Map