Assay Details
| Gene Symbol: PP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111974.4 | NP_187748.1 | 3 - 4 | 1083 | 67 | |
| GenBank mRNA | AY074368.1 | - | 3 - 4 | 1023 | 67 | |
| AY091391.1 | - | 3 - 4 | 882 | 67 | ||
| D38109.1 | - | 3 - 4 | 959 | 67 | ||
More Information
Set Membership: |
|

PP2CA
Genomic Map