Assay Details
| Gene Symbol: GPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111976.4 | NP_187750.1 | 1 - 2 | 807 | 63 | |
| GenBank mRNA | AK117634.1 | - | 1 - 2 | 629 | 63 | |
| BT005134.1 | - | 1 - 2 | 590 | 63 | ||
More Information
Set Membership: |
|

GPAT5
Genomic Map