Assay Details
| Gene Symbol: MYB65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337926.1 | NP_001327044.1 | 3 - 4 | 1630 | 75 | |
| NM_001337927.1 | NP_001327042.1 | 2 - 3 | 1556 | 75 | ||
| NM_111977.5 | NP_187751.1 | 3 - 4 | 1619 | 75 | ||
| GenBank mRNA | AY519585.1 | - | 2 - 3 | 1384 | 75 | |
| BT003940.1 | - | 3 - 4 | 1543 | 75 | ||
| BT005120.1 | - | 2 - 3 | 1384 | 75 | ||
More Information
Set Membership: |
|

MYB65
Genomic Map