Assay Details
| Gene Symbol: CYCB1;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337935.1 | NP_001326714.1 | 1 - 2 | 171 | 77 | |
| NM_111985.5 | NP_187759.2 | 1 - 2 | 171 | 77 | ||
| GenBank mRNA | AK227951.1 | - | 1 - 2 | 128 | 77 | |
| BT006437.1 | - | 1 - 2 | 37 | 77 | ||
More Information
Set Membership: |
|

CYCB1;3
Genomic Map