Assay Details
| Gene Symbol: CYCB1;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337935.1 | NP_001326714.1 | 3 - 4 | 648 | 98 | |
| NM_111985.5 | NP_187759.2 | 3 - 4 | 648 | 98 | ||
| GenBank mRNA | AK227951.1 | - | 3 - 4 | 605 | 98 | |
| BT006437.1 | - | 3 - 4 | 514 | 98 | ||
More Information
Set Membership: |
|

CYCB1;3
Genomic Map