Assay Details
| Gene Symbol: NHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111997.3 | NP_566395.1 | 1 | 904 | 150 | |
| GenBank mRNA | BT024621.1 | - | 1 | 611 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NHL2| Gene Symbol: NHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111997.3 | NP_566395.1 | 1 | 904 | 150 | |
| GenBank mRNA | BT024621.1 | - | 1 | 611 | 150 | |
Set Membership: |
|