Assay Details
| Gene Symbol: ATN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113672.3 | NP_189393.1 | 1 - 2 | 719 | 111 | |
| GenBank mRNA | AY058062.1 | - | 1 - 2 | 631 | 111 | |
| AY090300.1 | - | 1 - 2 | 133 | 111 | ||
More Information
Set Membership: |
|

ATN1
Genomic Map