Assay Details
| Gene Symbol: RST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113683.3 | NP_189404.2 | 14 - 15 | 3009 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RST1| Gene Symbol: RST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113683.3 | NP_189404.2 | 14 - 15 | 3009 | 111 | |
Set Membership: |
|