Assay Details
| Gene Symbol: MYB118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113694.2 | NP_189416.2 | 2 - 3 | 815 | 112 | |
| GenBank mRNA | DQ446708.1 | - | 2 - 3 | 593 | 112 | |
More Information
Set Membership: |
|

MYB118
Genomic Map