Assay Details
| Gene Symbol: DEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338921.1 | NP_001325789.1 | 1 - 2 | 505 | 75 | |
| NM_113709.5 | NP_189431.2 | 1 - 2 | 574 | 75 | ||
| GenBank mRNA | AY039585.1 | - | 1 - 2 | 453 | 75 | |
| AY113073.1 | - | 1 - 2 | 425 | 75 | ||
More Information
Set Membership: |
|

DEG1
Genomic Map