Assay Details
| Gene Symbol: DEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338921.1 | NP_001325789.1 | 4 - 5 | 836 | 87 | |
| NM_113709.5 | NP_189431.2 | 4 - 5 | 905 | 87 | ||
| GenBank mRNA | AY039585.1 | - | 4 - 5 | 784 | 87 | |
| AY113073.1 | - | 4 - 5 | 756 | 87 | ||
More Information
Set Membership: |
|

DEG1
Genomic Map