Assay Details
| Gene Symbol: DEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338921.1 | NP_001325789.1 | 7 - 8 | 1324 | 66 | |
| NM_113709.5 | NP_189431.2 | 7 - 8 | 1393 | 66 | ||
| GenBank mRNA | AY039585.1 | - | 7 - 8 | 1272 | 66 | |
| AY113073.1 | - | 7 - 8 | 1244 | 66 | ||
More Information
Set Membership: |
|

DEG1
Genomic Map