Assay Details
| Gene Symbol: AT3G28220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113741.4 | NP_189462.1 | 1 - 2 | 579 | 82 | |
| GenBank mRNA | AF372976.1 | - | 1 - 2 | 353 | 82 | |
| AY133588.1 | - | 1 - 2 | 340 | 82 | ||
More Information
Set Membership: |
|

AT3G28220
Genomic Map