Assay Details
| Gene Symbol: HMG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113794.4 | NP_189515.1 | 4 - 5 | 554 | 104 | |
| GenBank mRNA | BT002507.1 | - | 4 - 5 | 548 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMG| Gene Symbol: HMG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113794.4 | NP_189515.1 | 4 - 5 | 554 | 104 | |
| GenBank mRNA | BT002507.1 | - | 4 - 5 | 548 | 104 | |
Set Membership: |
|