Assay Details
| Gene Symbol: ABCB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113807.3 | NP_189528.1 | 4 - 5 | 979 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCB19| Gene Symbol: ABCB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113807.3 | NP_189528.1 | 4 - 5 | 979 | 106 | |
Set Membership: |
|