Assay Details
| Gene Symbol: ABCB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113807.3 | NP_189528.1 | 8 - 9 | 2594 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCB19| Gene Symbol: ABCB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113807.3 | NP_189528.1 | 8 - 9 | 2594 | 73 | |
Set Membership: |
|